Delmhorst 21-E/001 Moisture Meter Electrode, Extra-Deep Penetration/EIFS
Representative image only.
Delmhorst 21-E/001 Moisture Meter Electrode, Extra-Deep Penetration/EIFS
Delmhorst | Mfr: 21-E/001| Item: 5982005
Configure your meter to your needs
₹31,200.00INR / Each
₹31,200.00INR / Each
Buy It Together
Current Item
Delmhorst 21-E/001 Moisture Meter Electrode, Extra-Deep Penetration/EIFS
Delmhorst 21-E/001 Moisture Meter Electrode, Extra-Deep Penetration/EIFS
5982005
₹31,200.00INR / Each
+
Delmhorst PC/KIT Moisture Meter Computer Interface Kit
5982045
₹47,000.00INR / Each
+ +
Delmhorst FX-2000W/PS Digital Hay Moisture Meter
5982316
₹132,000.00INR / Each
=
Total Price 304800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flexibility to choose the right combination of electrodes and accessories for your application
  • 3 month warranty
More About this Item
Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..
Designed to find hidden moisture behind stucco, drywall, or for deep surface testing. The electrode is also insulated with only the tips being exposed, providing the only way to measure moisture content at various depths of penetration. Supplied with 7” pins for 6-1/4” penetration (pin 608/001).
3 Month Warranty
Specifications
  • Measurement TypePin
  • DescriptionMoisture Meter Electrode, Extra-Deep Penetration/EIFS