Delmhorst 19-E Moisture Meter Electrode, Two 5" Long Blade-Type; 1/Pk

Representative image only.

Delmhorst 19-E Moisture Meter Electrode, Two 5" Long Blade-Type; 1/Pk

Delmhorst | Mfr: 19-E| Item: 5982047

Configure your meter to your needs

₹49,100.00INR / Each

₹49,100.00INR / Each

Buy It Together

Current Item

Delmhorst 19-E Moisture Meter Electrode, Two 5" Long Blade-Type; 1/Pk
Delmhorst 19-E Moisture Meter Electrode, Two 5" Long Blade-Type; 1/Pk
5982047

₹49,100.00INR / Each
+
Delmhorst PC/KIT Moisture Meter Computer Interface Kit
5982045

₹47,000.00INR / Each
+ +
Delmhorst FX-2000W/PS Digital Hay Moisture Meter
5982316

₹132,000.00INR / Each
=
Total Price 322700.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Flexibility to choose the right combination of electrodes and accessories for your application
  • 3 month warranty

More About this Item

Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..

The two, 5” long blades allow for the measuring of moisture in between stacked paper materials.

3 Month Warranty
Specifications
  • Measurement TypePin
  • DescriptionMoisture Meter Electrode, Two 5" Long Blade-Type; 1/Pk
  • Warranty3 month
Reviews
Q&A