Delmhorst 12-E Moisture Meter Electrode, Double Roller-Type; 1/Pk
Representative image only.
Delmhorst 12-E Moisture Meter Electrode, Double Roller-Type; 1/Pk
Delmhorst | Mfr: 12-E| Item: 5982046
Configure your meter to your needs
₹98,300.00INR / Each
₹98,300.00INR / Each
Buy It Together
Current Item
Delmhorst 12-E Moisture Meter Electrode, Double Roller-Type; 1/Pk
Delmhorst 12-E Moisture Meter Electrode, Double Roller-Type; 1/Pk
5982046
₹98,300.00INR / Each
+
Delmhorst PC/KIT Moisture Meter Computer Interface Kit
5982045
₹47,000.00INR / Each
+ +
Delmhorst FX-2000W/PS Digital Hay Moisture Meter
5982316
₹132,000.00INR / Each
=
Total Price 371900.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flexibility to choose the right combination of electrodes and accessories for your application
  • 3 month warranty
More About this Item
Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..
Double roller type electrode are used for measuring moisture on a moving web or in textile.
3 Month Warranty
Specifications
  • Measurement TypeRoller
  • DescriptionMoisture Meter Electrode, Double Roller-Type; 1/Pk