CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Male Luer, Polycarbonate

Representative image only.

CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Male Luer, Polycarbonate

CPC (Colder) | Mfr: CNX170LMHT| Item: 5013826

Micro-flow genderless sterile connectors simplify process integration while maximizing flexibility

₹5,400.00INR / Each

₹5,400.00INR / Each

Buy It Together

Current Item

CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Male Luer, Polycarbonate
CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Male Luer, Polycarbonate
5013826

₹5,400.00INR / Each
+ + + +
Cole-Parmer PVC 2-Stop Pump Tubing, 0.76 mm, Black/Black; 12/Pk
5013725

₹4,700.00INR / Pkg of 12
+ =
Total Price 173100.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Simple “pinch, click, and pull” design minimizes operator errors
  • Simplified tubing connection provides high performance and reliability
  • Designed for micro-flow media transfer applications
  • Free technical application support available to determine which connector type is needed for your application

More About this Item

The MicroCNX® genderless luer connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing a sterile, closed connection on an existing luer connection. Connectors eliminate the need for tube welding, saving you time and money. To use, simply pinch to remove the protective cover, join two halves together and squeeze until you hear an audible “click”, then pull the membrane strips away from the connector.

These aseptic quick connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Seal MaterialPlatinum-Cured Silicone
  • Sterilization MethodGamma irradiation up to 50 kGy or autoclave one cycle up to 130°C (266°F) for 60 minutes
  • Connection TypeQuick Disconnect x Male Luer
  • Flow PatternStraight
  • ColorWhite with teal protective cover
  • Min Temperature (° F)34
  • Max Temperature (° F)104
  • Min Temperature (° C)1
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)43.5
  • Fitting Length (in)2 5/32
  • Fitting Width (in)1 31/64
  • Fitting Depth (in)59/64
  • Fitting Length (mm)54.6
  • Fitting Width (mm)37.8
  • Fitting Depth (mm)23.6
  • Regulatory ApprovalsFDA;USP;Non-DEHP;Non-Phthalate;ADCF;RoHS
  • Luer Connection Type (Port 1)Male
  • DescriptionQuick-Disconnect Fitting, Micro-Flow, Genderless Male Luer, Polycarbonate
  • WarrantyManufacturer defect
Reviews
Q&A