PharmaLok Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 0.5” (12.7 mm) to 0.75” (19.0 mm)

Representative image only.

PharmaLok Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 0.5” (12.7 mm) to 0.75” (19.0 mm)

| Mfr: CPUN075-U71| Item: 5013353

Precision 360° sealing for reliable, tool-free biopharma connections

₹2,900.00INR / Each

₹2,900.00INR / Each

Buy It Together

Current Item

PharmaLok Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 0.5” (12.7 mm) to 0.75” (19.0 mm)
PharmaLok Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 0.5” (12.7 mm) to 0.75” (19.0 mm)
5013353

₹2,900.00INR / Each
+ + + + + +
Cole-Parmer PVC 2-Stop Pump Tubing, 0.76 mm, Black/Black; 12/Pk
5013725

₹4,700.00INR / Pkg of 12
+ =
Total Price 181900.00 ADD 8 ITEMS TO CART
Product Description

Key Features

  • Universal compatibility supports a wide range of fittings and flanges
  • Adaptive closure accommodates gasket and flange variations
  • Easy-to-use, one-handed closure - no tools required
  • Compact design with nested latch saves space and eliminates protrusions
  • Clean assembly minimizes particulate
  • Connects both plastic and stainless steel fittings
  • Sterilize by autoclave, gamma irradiation, or x-ray
  • ASME BPE 018 DT-7-1 and SIP/CIP compatible
  • Manufacturer defect warranty

More About this Item

These hygienic union TC sanitary clamps offer 360° uniform compression for consistent, secure sealing without gasket damage. Unlike single-hinge clamps, their innovative design evenly distributes pressure, avoiding pinching or distortion. With universal fit, tool-free operation, and compatibility across plastic and stainless-steel fittings, they simplify biopharma workflows. The adaptive closure handles gasket and flange variations, and the compact design uses up to 25% less material. Built to withstand over 100 psi, they’re ideal for demanding biopharma applications.

The clamps are engineered for sterilization, with compatibility for autoclaving up to 273°F (134°C) for 30 minutes, as well as gamma and X-ray sterilization up to 50 kGy.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Specifications
  • Clamp MaterialNylon
  • Connector MaterialNylon
  • Clamp TypeHygienic union TC sanitary clamp
  • ColorWhite body, blue lever
  • Diameter (in)0.75
  • DescriptionHygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 0.5” (12.7 mm) to 0.75” (19.0 mm)
  • WarrantyManufacturer defect
Reviews
Q&A