PharmaLok™ Hygienic Union TC Clamps
Precision 360° sealing for reliable, tool-free biopharma connections
  • Universal compatibility supports a wide range of fittings and flanges
  • Adaptive closure accommodates gasket and flange variations
  • Easy-to-use, one-handed closure - no tools required
  • Compact design with nested latch saves space and eliminates protrusions
  • Clean assembly minimizes particulate
  • Connects both plastic and stainless steel fittings
  • Sterilize by autoclave, gamma irradiation, or x-ray
  • ASME BPE 018 DT-7-1 and SIP/CIP compatible
These hygienic union TC sanitary clamps offer 360° uniform compression for consistent, secure sealing without gasket damage. Unlike single-hinge clamps, their innovative design evenly distributes pressure, avoiding pinching or distortion. With universal fit, tool-free operation, and compatibility across plastic and stainless-steel fittings, they simplify biopharma workflows. The adaptive closure handles gasket and flange variations, and the compact design uses up to 25% less material. Built to withstand over 100 psi, they’re ideal for demanding biopharma applications.
The clamps are engineered for sterilization, with compatibility for autoclaving up to 273°F (134°C) for 30 minutes, as well as gamma and X-ray sterilization up to 50 kGy.
2,900.00 - ₹3,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Diameter (in) Description
Availability Pricing
PharmaLok Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 0.5” (12.7 mm) to 0.75” (19.0 mm) 5013353 Mfr # CPUN075-U71
COMPARE
Nylon 0.75 Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 0.5” (12.7 mm) to 0.75” (19.0 mm)
₹2,900.00INR / Each
PharmaLok Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue Lever, 1" (25.4 mm) to 1.5” (38.1 mm) 5013354 Mfr # CPUX150-U71
COMPARE
Nylon 1.5 Hygienic Union TC Clamp, Glass-Reinforced White Nylon Blue, Lever, 1" (25.4 mm) to 1.5” (38.1 mm)
₹3,000.00INR / Each