CPC (Colder) MicroCNX® Nano Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8” ID

Representative image only.

CPC (Colder) MicroCNX® Nano Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8” ID

CPC (Colder) | Mfr: CNY17102HT| Item: 5013831

Micro-flow nano connectors simplify sterile tubing connections for small-format bioprocessing

₹6,300.00INR / Each

₹6,300.00INR / Each

Buy It Together

Current Item

CPC (Colder) MicroCNX® Nano Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8” ID
CPC (Colder) MicroCNX® Nano Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8” ID
5013831

₹6,300.00INR / Each
+ + +
Cole-Parmer PVC 2-Stop Pump Tubing, 1.85 mm, Green/Green; 12/Pk
5013755

₹4,700.00INR / Pkg of 12
+ =
Total Price 37100.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Withstand ultra-low temperatures down to -190°C
  • Simple “pinch, click, and pull” design—no tools required
  • Enables fast and error-free connections
  • Tailored for cell and gene therapy (CGT)
  • Designed for micro-flow media transfer applications
  • Free technical application support available to determine which connector type is needed for your application
  • 1/8" hose barb

More About this Item

The MicroCNX® genderless nano connectors offer a fast, simple, and secure solution for connecting tubing in small-format biomanufacturing assemblies. Available with 1/8” and 1/16” hose barb terminations, they are compatible with a wide range of tubing materials including silicone, TPE, and PVC. Engineered to withstand ultra-low temperatures down to -190°C, the connectors are ideal for integration with freezing cassettes used in cell and gene therapy (CGT) processing.

Designed for aseptic operation, the connectors maintain a closed flow path—ensuring sterile connections even in non-sterile environments. They eliminate the need for tube welding, helping reduce setup time and overall costs. Operation is intuitive: pinch to remove the protective cover, align and join the two halves, then squeeze until you hear an audible "click." Finally, pull away the membrane strips to complete the sterile connection.

The connectors are purpose-built for secure, sterile fluid transfer in CGT and other biological media applications.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Fitting MaterialPolyphenylsulfone
  • Seal MaterialPlatinum-Cured Silicone
  • Sterilization MethodGamma irradiation up to 50 kGy or autoclave one cycle up to 130°C (266°F) for 60 minutes
  • Connection TypeQuick Disconnect
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Flow PatternStraight
  • ColorOff white with gold protective cover
  • Min Temperature (° F)34
  • Max Temperature (° F)104
  • Min Temperature (° C)1
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)29
  • Fitting Length (in)2 5/32
  • Fitting Width (in)1 7/16
  • Fitting Depth (in)5/16
  • Fitting Length (mm)54.9
  • Fitting Width (mm)36.6
  • Fitting Depth (mm)7.9
  • Regulatory ApprovalsRoHS;ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • DescriptionULT Nano Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8” ID
  • WarrantyManufacturer defect
Reviews
Q&A