COMARK PK23M Thermocouple Oven Meat Probe, Type K, Stainless Steel
Representative image only.
COMARK PK23M Thermocouple Oven Meat Probe, Type K, Stainless Steel
COMARK | Mfr: PK23M| Item: 9002588
Designed specifically for use in the food industry
₹22,300.00INR / Each
for an additional
₹30,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹22,300.00INR / Each
Buy It Together
Current Item
COMARK PK23M Thermocouple Oven Meat Probe, Type K, Stainless Steel
COMARK PK23M Thermocouple Oven Meat Probe, Type K, Stainless Steel
9002588
₹22,300.00INR / Each
+ =
Total Price 65000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Most probes have a response time of under 0.5 seconds
  • Include a 7-foot coil cord ( except for the ATT29 probe which is 4-foot cord )
  • Measures meat and food temperatures during cooking up to 482°F (250°C)
  • Fitted with 2-pin sub-miniature connector
  • 6 month warranty
More About this Item
Comark food probes are designed specifically for use in the food industry to ensure proper food preparation. Penetration probes are designed to be inserted directly into the food, typically poultry, beef or pork, to gauge the internal temperature and to ensure that the meat is cooked properly for safe eating. Surface probes are used to make sure the cooking surface is at the desired temperature before and during the cooking process.
This thermocouple probe is ideal for checking meat and food temperatures during cooking. The Type K probe is suitable for use with a range of Comark thermometers.
6 Month Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-58
  • Max Temperature (° F)482
  • Min Temperature (° C)-50
  • Max Temperature (° C)250
  • Response Time (Sec)2
  • Connector TypeSub-mini (2-pin)
  • Cable TypeCoiled
  • DescriptionThermocouple Oven Meat Probe, Type K, Stainless Steel