Cole-Parmer Teasing Needle with Insulated Handle, 6", Straight, 19 Gauge
Representative image only.
Cole-Parmer Teasing Needle with Insulated Handle, 6", Straight, 19 Gauge
Cole-Parmer | Item: 0439647
Nichrome wire inoculating needles for selecting, inoculating, or transferring microbiological samples
₹4,400.00INR / Each
₹4,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Teasing Needle with Insulated Handle, 6", Straight, 19 Gauge
Cole-Parmer Teasing Needle with Insulated Handle, 6", Straight, 19 Gauge
0439647
₹4,400.00INR / Each
+ + + + =
Total Price 57500.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Cools quickly after sterilization
  • Less expensive than platinum
  • 6” straight needle
  • Insulated, light-weight aluminum handle protects hand during use
More About this Item
These inoculating needles are made from high quality nichrome wire that cools rapidly after sterilization. They are more economical than platinum needles. Available with or without handle and straight or angled needle.
Specifications
  • SterileNo
  • B and S Wire Gauge19
  • DescriptionTeasing Needle with Insulated Handle, 6", Straight, 19 Gauge