Cole-Parmer® Nichrome Inoculating Loops without Handle
Reusable inoculating loops used to select, inoculate, or transfer microbiological samples
  • Glides over agar surface smoothly without damage
  • Nichrome wire cools quickly after sterilization
  • Less expensive than platinum loops
  • Use with your favorite handle
Used to select, transfer and inoculate living microorganisms in the streaking process, these inoculating loops have been designed to slide easily along agar surfaces without cutting or tearing. Made of 26-gauge twisted nichrome wire, they cool rapidly and are more economical than platinum loops.
Supplied without handles, these non-sterile, reusable inoculating loops are available calibrated or non-calibrated. Brass handles are sold separately as items 04396-40 and 04396-41.
11,200.00 - ₹42,500.00INR / Pkg of 5
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
B and S Wire Gauge Loop Inside Diameter (mm) Description
Availability Pricing
COMPARE
26 4 Nichrome Inoculating Loops without Handle, Calibrated, 10 uL, 4 mm ID; 5/PK
₹29,300.00INR / Pkg of 5
COMPARE
26 1.45 Nichrome Inoculating Loops without Handle, Calibrated, 1 uL, 1.45 mm ID; 5/PK
₹42,500.00INR / Pkg of 5
COMPARE
26 2.45 Nichrome Inoculating Loops without Handle, Small, 2.45 mm ID; 25/PK
₹12,900.00INR / Pkg of 25
COMPARE
26 3.41 Nichrome Inoculating Loops without Handle, Medium, 3.41 mm ID; 25/PK
₹11,200.00INR / Pkg of 25
COMPARE
26 5.45 Nichrome Inoculating Loops without Handle, Large, 5.45 mm ID; 25/PK
₹11,200.00INR / Pkg of 25