Cole-Parmer® Inoculating Loop and Needle Holders, Brass
Brass holder safely and securely holds inoculating loops or needles
  • Helps prevent direct contact with sample
  • Insulated to protect hand during use
These reusable brass handles securely hold inoculating loops and needles allowing you to select, inoculate or transfer microbiological samples during the streaking process. They are insulated to protect the hand during sterilization. Loops and needles are sold separately.
6,700.00 - ₹9,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Handle Type Overall Length (mm) Description
Availability Pricing
COMPARE
Brass 152.4 Inoculating Loop and Needle Holder, 15.24 cm (6 in), Brass
₹6,700.00INR / Each
COMPARE
Brass 203.2 Inoculating Loop and Needle Holder, 20.32 cm (8 in), Brass
₹9,100.00INR / Each