Cole-Parmer Spill Tray for 48" Fume Hood, Black Polypropylene

Representative image only.

Cole-Parmer Spill Tray for 48" Fume Hood, Black Polypropylene

Cole-Parmer | Item: 7890011

Accessories for your Cole-Parmer ductless fume hood

₹84,100.00INR / Each

₹84,100.00INR / Each

Buy It Together

Current Item

Cole-Parmer   Spill Tray for 48" Fume Hood, Black Polypropylene
Cole-Parmer Spill Tray for 48" Fume Hood, Black Polypropylene
7890011

₹84,100.00INR / Each
+
Chemical Resistant Tray - 13-1/4" X 10-5/8", 1 Ea
0671580

₹7,400.00INR / Each
+ + + + + =
Total Price 3373900.00 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Constructed from chemically resistant polypropylene
  • Compatible with 48" Cole-Parmer Ductless Fume Hood, 78900-04 & 78900-05
  • 1 year warranty

More About this Item

Choose from a wide range of accessories for enhanced performance

1 Year Warranty
Specifications
  • MaterialPolypropylene
  • ColorBlack
  • Compatible With48" Fume hood
  • Width (InchShort)48
  • Width (cm)121.9
  • DescriptionSpill Tray for 48" Fume Hood, Black Polypropylene
  • Warranty1 Year
Reviews
Q&A