Cole-Parmer SamplePrep Foam Cryo Holder
Representative image only.
Cole-Parmer SamplePrep Foam Cryo Holder
Cole-Parmer SamplePrep | Mfr: 2266| Item: 6121006
Securely hold vials in place while processing samples
₹18,000.00INR / Each
₹18,000.00INR / Each
Product Description
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Securely hold vials in place while processing samples
  • Safely grind samples
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.
Holds two 2666 (2.0 mL micro-centrifuge tubes) cryo-blocks. Holder allows cryo-blocks to be held secure, lifted easily in and out of the 2010 Geno/Grinder clamp assembly. Cryo-blocks sold separately.
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.
Specifications
  • Tubes Held96
  • MaterialFoam
  • DescriptionFoam Cryo Holder