Cole-Parmer SamplePrep 2100 Grinding Ball Dispenser for 96-Well Microplates; 1/EA
Representative image only.
Cole-Parmer SamplePrep 2100 Grinding Ball Dispenser for 96-Well Microplates; 1/EA
Cole-Parmer SamplePrep | Mfr: 2100| Item: 0450018
Get the most out of your Cole-Parmer® homogenizer
₹44,600.00INR / Each
₹44,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer SamplePrep 2100 Grinding Ball Dispenser for 96-Well Microplates; 1/EA
Cole-Parmer SamplePrep 2100 Grinding Ball Dispenser for 96-Well Microplates; 1/EA
0450018
₹44,600.00INR / Each
+ + + + + + =
Total Price 346000.00 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Choose from a wide range of accessories for your homogenizer
  • Loads an entire 96-well microplate with grinding balls in less than a second
  • Compatible with 5/32” (4 mm) grinding balls 04500-19
  • Compatible with standard 96-well microplates
More About this Item
Optimize your homogenizer with these quality accessories.
The dispenser simultaneously dispenses one 5/23” (4 mm) steel grinding ball into Eash well of a standard 96-well microplate.
Specifications
  • Use with Generators96-well microplate and grinding ball 04500-19
  • DescriptionGrinding Ball Dispenser for 96-Well Microplates; 1/EA