Cole-Parmer Safety Wash Bottle, LDPE, Vented, Methanol, 500mL (16oz); 6/PK

Representative image only.

Cole-Parmer Safety Wash Bottle, LDPE, Vented, Methanol, 500mL (16oz); 6/PK

Cole-Parmer | Item: 6230026

Built-in vent releases built-up pressure, prevents leaking

₹8,800.00INR / Pkg of 6

₹8,800.00INR / Pkg of 6

Buy It Together

Current Item

Cole-Parmer Safety Wash Bottle, LDPE, Vented, Methanol, 500mL (16oz); 6/PK
Cole-Parmer Safety Wash Bottle, LDPE, Vented, Methanol, 500mL (16oz); 6/PK
6230026

₹8,800.00INR / Pkg of 6
+ +
Dynalon Polypropylene (PP) Paddled Stir Rod, 9-3/4" L; 10/Pk
0637300

₹6,400.00INR / Pkg of 10
+
Cole-Parmer Wide-Mouth Plastic Bottle, HDPE, 4000mL
0605520

₹6,200.00INR / Each
=
Total Price 27500.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Silk-screened labels resist chemicals, won’t wear off
  • Easy identification with color coded closures
  • 1 year warranty

More About this Item

These self venting wash bottles display the chemical name, formula, NFPS hazard rating, DOT symbols, and the Chemical Abstract Service (CAS). Valve mechanism automatically vents volatile solvent vapor and eliminates potentially hazardous problems of solvent drip and sputter. Angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow. A wide mouth makes bottles easy to fill. View contents through translucent bottle; easily identify contents by color-coded closures. The closure and stem are molded as one piece to make them leak-proof.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

1 Year Warranty
Specifications
  • Capacity (mL)500
  • Capacity (L)0.5
  • Capacity (oz)17
  • MaterialLow Density Polyethyene (LDPE)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size53 mm
  • Cap ColorGreen
  • Bottle ColorTranslucent
  • AutoclavableNo
  • Label TypeMethanol
  • Neck Inside Diameter (ID) (mm)42.4
  • Neck Inside Diameter (ID) (in)1 43/64
  • Outside Diameter (OD) (in)2 27/32
  • Outside Diameter (OD) (cm)7.23
  • Overall Height (in)6 31/64
  • Overall Height (cm)16.5
  • DescriptionSafety Wash Bottle, LDPE, Vented, Methanol, 500mL; 6/PK
  • Warranty1 year
Reviews
Q&A