Cole-Parmer® Safety Wash Bottles, LDPE, Vented
Built-in vent releases built-up pressure, prevents leaking
  • Silk-screened labels resist chemicals, won’t wear off
  • Easy identification with color coded closures
These self venting wash bottles display the chemical name, formula, NFPS hazard rating, DOT symbols, and the Chemical Abstract Service (CAS). Valve mechanism automatically vents volatile solvent vapor and eliminates potentially hazardous problems of solvent drip and sputter. Angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow. A wide mouth makes bottles easy to fill. View contents through translucent bottle; easily identify contents by color-coded closures. The closure and stem are molded as one piece to make them leak-proof.
5,800.00 - ₹12,000.00INR / Pkg of 6
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Label Type Material
Availability Pricing
COMPARE
500 Acetone Low Density Polyethyene (LDPE)
₹11,000.00INR / Pkg of 6
COMPARE
500 Ethanol Low Density Polyethyene (LDPE)
₹12,000.00INR / Pkg of 6
COMPARE
1000 Ethanol Low Density Polyethyene (LDPE)
₹5,800.00INR / Pkg of 4
COMPARE
500 Methanol Low Density Polyethyene (LDPE)
₹6,300.00INR / Pkg of 6
COMPARE
1000 Methanol Low Density Polyethyene (LDPE)
₹5,800.00INR / Pkg of 4
COMPARE
1000 Acetone Low Density Polyethyene (LDPE)
₹6,000.00INR / Pkg of 4
COMPARE
500 Methyl Ethyl Ketone Low Density Polyethyene (LDPE)
₹6,500.00INR / Pkg of 6
COMPARE
500 Sodium Hypochlorite Low Density Polyethyene (LDPE)
₹5,900.00INR / Pkg of 6
COMPARE
500 Isopropanol Low Density Polyethyene (LDPE)
₹6,300.00INR / Pkg of 6
COMPARE
1000 Isopropanol Low Density Polyethyene (LDPE)
₹6,200.00INR / Pkg of 4
COMPARE
500 Distilled Water Low Density Polyethyene (LDPE)
₹6,300.00INR / Pkg of 6
COMPARE
1000 Distilled Water Low Density Polyethyene (LDPE)
₹5,800.00INR / Pkg of 4
COMPARE
500 Assorted Low Density Polyethyene (LDPE)
₹6,300.00INR / Pkg of 6
COMPARE
500 Self Labeling Bottle Low Density Polyethyene (LDPE)
₹10,000.00INR / Pkg of 6
Showing 1- 14 of 14