Cole-Parmer Removable Double-Junction ORP Electrode
Representative image only.
Cole-Parmer Removable Double-Junction ORP Electrode
Cole-Parmer | Item: 2700931
Easily remove electrodes in seconds without removing the holder
₹44,100.00INR / Each
₹44,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer Removable Double-Junction ORP Electrode
Cole-Parmer Removable Double-Junction ORP Electrode
2700931
₹44,100.00INR / Each
+ =
Total Price 56200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
More About this Item
Use for oxidation/reduction measurements in plating baths, waste-water monitoring, and other general-purpose applications. Platinum band ORP withstands temperatures from 23 to 212ºF (-5 to 100ºC).
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • ConnectorBNC
  • DescriptionRemovable Double-Junction ORP Electrode