Cole-Parmer Electrode Holder for Removable pH and ORP Electrodes, CPVC
Representative image only.
Cole-Parmer Electrode Holder for Removable pH and ORP Electrodes, CPVC
Cole-Parmer | Item: 2700950
Easily remove electrodes in seconds without removing the holder
₹12,100.00INR / Each
₹12,100.00INR / Each
Product Description
Key Features
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
More About this Item
Integral cap assembly permits quick servicing for calibrations or replacement. Electrode holders have threads on each end. Install holders into a pipe tee for in-line use or on the end of a submersion pipe. Choose from CPVC for economical aplications or PVDF for enhanced chemical resistance. Electrodes order separately.
1 Year Warranty
Specifications
  • Electrode Body MaterialCPVC
  • Min Temperature (° C)-5
  • Max Temperature (° C)80
  • Max Pressure (PSI)100
  • Thread Size NPT(M) (in)3/4
  • DescriptionElectrode Holder for Removable pH and ORP Electrodes, CPVC