Cole-Parmer® ORP Electrodes with Integrated Assemblies
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
Use for oxidation/reduction measurements in plating baths, waste-water monitoring, and other general-purpose applications. Platinum band ORP withstands temperatures from 23 to 212ºF (-5 to 100ºC).
35,700.00 - ₹44,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Removable Single-Junction ORP Electrode
₹35,700.00INR / Each
COMPARE
Removable Double-Junction ORP Electrode
₹44,100.00INR / Each