Cole-Parmer Open Ended Pipettes, 2 mL, Individually Wrap, Graduated, Sterile; 500/Cs
Representative image only.
Cole-Parmer Open Ended Pipettes, 2 mL, Individually Wrap, Graduated, Sterile; 500/Cs
Cole-Parmer | Item: 0439514
Eliminate clogged tips
₹10,000.00INR / Case of 500
₹10,000.00INR / Case of 500
Buy It Together
Current Item
Cole-Parmer Open Ended Pipettes, 2 mL, Individually Wrap, Graduated, Sterile; 500/Cs
Cole-Parmer Open Ended Pipettes, 2 mL, Individually Wrap, Graduated, Sterile; 500/Cs
0439514
₹10,000.00INR / Case of 500
+ + + =
Total Price 49200.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Made from transparent medical grade polystyrene (PS) sterilized by gamma irradiation
  • Each lot is tested for endotoxins, cytotoxicity, and are guaranteed nonhemolytic
More About this Item
Serological pipettes feature a wide end opening instead of a tip, making them ideal for pipetting thick or viscous liquids containing particulates which could block a standard tip. The pipettes are also beneficial in applications where speed is critical. Pipettes come individually wrapped.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • SterileYes
  • Max Volume (mL)2
  • Graduation Interval0.01
  • Body MaterialPolystyrene
  • Color CodeWhite
  • DescriptionOpen Ended Pipettes, 2 mL, Individually Wrap, Graduated, Sterile; 500/Cs