Cole-Parmer® Open-End Serological Pipettes
Eliminate clogged tips
  • Made from transparent medical grade polystyrene (PS) sterilized by gamma irradiation
  • Each lot is tested for endotoxins, cytotoxicity, and are guaranteed nonhemolytic
Serological pipettes feature a wide end opening instead of a tip, making them ideal for pipetting thick or viscous liquids containing particulates which could block a standard tip. The pipettes are also beneficial in applications where speed is critical. Pipettes come individually wrapped.
5,900.00 - ₹10,000.00INR / Case of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL)
Availability Pricing
COMPARE
1
₹8,500.00INR / Case of 500
COMPARE
2
₹10,000.00INR / Case of 500
COMPARE
5
₹5,900.00INR / Case of 200
COMPARE
10
₹8,900.00INR / Case of 200