Cole-Parmer Nitrate (NO3-) ISE Double junction solution kits
Representative image only.
Cole-Parmer Nitrate (NO3-) ISE Double junction solution kits
Cole-Parmer | Item: 2750279
Solutions needed for accurate measurements
₹24,500.00INR / Each
₹24,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer Nitrate (NO3-) ISE Double junction solution kits
Cole-Parmer Nitrate (NO3-) ISE Double junction solution kits
2750279
₹24,500.00INR / Each
+
Cole-Parmer Replacement ionic strength adjustor, TISAB I, 1 gallon
2750358
₹29,400.00INR / Gallon of 1
+
Cole-Parmer ISE Standard Solution, Silver; 1000 ppm
2750332
₹11,000.00INR / Each
+ +
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
₹64,600.00INR / Each
+ +
Cole-Parmer ISE Electrode; Ammonium
2707701
₹68,300.00INR / Each
+ + + =
Total Price 383500.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Multiple ion solutions are available
  • 1 year warranty
More About this Item
Choose the compatible solution kit to operate your Cole-Parmer ion-selective electrodes properly. Kits contain a 60 mL bottle of fill solution, a 475 mL bottle of standard, and a 475 mL bottle of ISA.
Individual bottles also listed.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction, membrane
  • Reference Electrolyte0.1 M (NH4)2SO4 with 0.01 M KCl/1000 ppm as NO3-
  • Ion-Selective ElectrodeNitrate
  • DescriptionNitrate (NO3-) ISE Double junction solution kits