Cole-Parmer Nickel/SS Lab Spatulas with Flat Ends; 3/PK
Representative image only.
Cole-Parmer Nickel/SS Lab Spatulas with Flat Ends; 3/PK
Cole-Parmer | Item: 0636905
Get precise sampling of powders or salts
₹3,300.00INR / Pkg of 3
₹3,300.00INR / Pkg of 3
Buy It Together
Current Item
Cole-Parmer Nickel/SS Lab Spatulas with Flat Ends; 3/PK
Cole-Parmer Nickel/SS Lab Spatulas with Flat Ends; 3/PK
0636905
₹3,300.00INR / Pkg of 3
+ + +
Cole-Parmer Spatula Macro Spoon, Stainless Steel,120mm; 3/PK
0628704
₹1,900.00INR / Pkg of 3
+ + + + + =
Total Price 74100.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easy-to-clean finish
  • Choose the right end style for your needs
More About this Item
Nickel/stainless steel spatulas come in a variety of end styles. All spatulas have a glazed finish, except for models 06369-05, and -16 which have a mirror finish.
Specifications
  • Overall Length (in)8 1/4
  • Overall Length (cm)20.96
  • Material304 Stainless Steel, Nickel
  • SterileNo
  • DescriptionNickel/SS Lab Spatulas with Flat Ends; 3/PK