Glass Stir Rods
Manufactured with borosilicate glass with rounded annealed ends. Stir rods are highly chemical resistant and withstands temperatures to 400° F (205°)C. Autoclavable.
5,700.00 - ₹45,300.00INR / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in)
Availability Pricing
DWK Life Sciences (Kimble) Stir Rod, Glass, 5 mm dia X 150 mm L, 25/pk WW-06369-21 Mfr # 40500-150
COMPARE
Glass 6
₹5,700.00INR / Pkg of 25
DWK Life Sciences (Kimble) Stir Rod, Glass, 6.4 mm dia X 250 mm L, 25/pk WW-06369-23 Mfr # 40500-250
COMPARE
Glass 9 7/8
₹14,600.00INR / Pkg of 25
DWK Life Sciences (Kimble) Stir Rod, Glass, 9.5 mm dia X 375 mm L, 25/pk WW-06369-25 Mfr # 40500-375
COMPARE
Glass 14 3/4
₹45,300.00INR / Pkg of 25