Cole-Parmer Imhoff Settling Cones and Rack Kit; Three Cones, One Rack
Representative image only.
Cole-Parmer Imhoff Settling Cones and Rack Kit; Three Cones, One Rack
Cole-Parmer | Item: 2500028
Bundled for convenience and to save you money!
₹74,200.00INR / Each
₹74,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Imhoff Settling Cones and Rack Kit; Three Cones, One Rack
Cole-Parmer Imhoff Settling Cones and Rack Kit; Three Cones, One Rack
2500028
₹74,200.00INR / Each
+ =
Total Price 137800.00 ADD 2 ITEMS TO CART
Product Description
Includes
three settling cones and one heavy-gauge wire rack.
Key Features
  • Three cones, one rack
  • Use settling cones and rack to determine settleable materials in water and wastewater samples, and sand in well water
  • Cones are externally graduated in mL from the bottom upward: lines at 0.3 and 0.4 mL, 0.5 to 1 mL in 0.1 mL increments, 1 to 10 mL in 0.5 mL increments, 10 to 40 mL in 1 mL increments, 40 to 100 mL in 2 mL increments, and then lines at 250, 500 and 1000 mL
More About this Item
The reusable, 1-L Imhoff settling cones are made of clear styrene-acrylonitrile and feature a leakproof, screw-type plug. The heavy-duty wire rack can hold all three cones at once and measures 17-1/2"L x 5-1/2"W x 16"H.
Specifications
  • DescriptionImhoff Settling Cones and Rack Kit; Three Cones, One Rack