Cole-Parmer Handheld Tubing Cutter, PTFE Coated Steel, Max 1-1/4" OD Tubing
Representative image only.
Cole-Parmer Handheld Tubing Cutter, PTFE Coated Steel, Max 1-1/4" OD Tubing
Cole-Parmer | Item: 5011850
Durable steel blade makes straight, clean cuts through plastic tubing
₹2,100.00INR / Each
₹2,100.00INR / Each
Product Description
Key Features
  • Steel blade offers durability
  • Cuts through tubing with OD up to 1-1/4" (up to 1-1/2" OD for polyethylene tubing)
More About this Item
Use tubing cutters to make clean, straight cuts through your plastic tubing. Straight cuts ensure better connection to fittings.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • MaterialPTFE-Coated Steel
  • ColorBlack
  • Compatible WithWork with tubing up to 1-1/4" OD (1-1/2" OD for PE tubing)
  • DescriptionHandheld Tubing Cutter, PTFE Coated Steel, Max 1-1/4 OD Tubing