Cole-Parmer FlexiRoll Digital Tube Roller Shaker; 220 VAC

Representative image only.

Cole-Parmer FlexiRoll Digital Tube Roller Shaker; 220 VAC

Cole-Parmer | Item: 0439745

Use with a variety of bottles, tubes, and flasks

₹376,000.00INR / Each

₹376,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer FlexiRoll Digital Tube Roller Shaker; 220 VAC
Cole-Parmer FlexiRoll Digital Tube Roller Shaker; 220 VAC
0439745

₹376,000.00INR / Each
+
Ibi Scientific Low-Profile Bottle Roller; 90-240 VAC, 50/60 Hz
3620203

₹1,099,000.00INR / Each
+ + +
Cole-Parmer SH-200D-R-L Large Reciprocating Shaker; 230 VAC
5190102

₹173,000.00INR / Each
=
Total Price 3590000.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Mix your tubes and bottles with minimal aeration
  • Double your capacity and save precious lab space by stacking two units
  • 1 year warranty

More About this Item

This compact digital roller shaker can be used with variety of bottles, tubes, and flasks. Safely operate on a benchtop or in an incubator from 4 to 60°C, and up to 95% humidity. Digitally set the rotation rate from 0.5 to 80 rpm.

Rollers may be added and removed for additional versatility to accept a wider range of bottle and tube sizes. If using on centrifuge and test tubes, O-rings are required for placement around the sample tubes to assure safe mixing. Models 24410-14 allows you to stack up to two rollers to double your capacity and save bench space. Order the optional remote controller to also obtain timed run functionality.

Ideal for growth and culturing of adherent cells, suspension cells, mixing test tubes, washing nucleic acid blots, or performing hybridization protocols. In Western, Southern, and Northern blotting applications, use as an alternative to the more expensive rotisserie style hybridization ovens. Purchase five additional rollers the FlexiRoll can rotate 20 to 15 and 50 mL tubes at one time, ensuring a complete and uniform mixing of the reagents with the filter paper.

Conformite Europeenne
Specifications
  • Degree of Rotation (degrees)360
  • Min Speed (rpm)0.5
  • Max Speed (rpm)80
  • CapacityDepends on set up and tubes/bottles used
  • Min Operating Temperature (° C)4
  • DisplayDigital
  • Power (VAC)240
  • Power (Hz)50
  • DescriptionDigital Tube Roller Shaker; 220 VAC
  • Warranty1 Year
Reviews
Q&A