Cole-Parmer Fixed-Volume Pipettor, 5 µL
Representative image only.
Cole-Parmer Fixed-Volume Pipettor, 5 µL
Cole-Parmer | Item: 2450111
Ergonomic finger grip for comfortable use
₹3,500.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹3,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer Fixed-Volume Pipettor, 5 µL
Cole-Parmer Fixed-Volume Pipettor, 5 µL
2450111
₹3,500.00INR / Each
+ +
Cole-Parmer Pipette Tip, 0.1 to 10 µL, Bulk; 1000/BG
2450142
₹4,500.00INR / Bag of 1000
+ +
Cole-Parmer EX+ Adjustable Volume Pipettor, 1-10 mL; Each
0791125
₹69,000.00INR / Each
+
Cole-Parmer EX Adjustable Volume Pipette, 0.1-2.0 uL; Each
0790901
₹66,600.00INR / Each
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
+
Cole-Parmer Pipette Tip, 100 to 1000 µL, Bulk; 1000/BG
2450261
₹5,700.00INR / Bag of 1000
=
Total Price 173500.00 ADD 8 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Color-coding drop and removal tool, calibration tool and silicon grease, along with sample tips and calibration report.
Key Features
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Volume: 5 µL
  • 3 year warranty
More About this Item
These pipettors are offered in a wide-range of fixed volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.
3 Year WarrantyConformite EuropeenneISO 9001:2008 Certified Supplier
Specifications
  • AutoclavableYes
  • Max Volume (µL)5
  • Accuracy (%)1.3
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeYellow
  • DescriptionFixed-Volume Pipettor, 5 µL; 1/EA