Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4" Clamp x 1/2" ID; 1/Each
Representative image only.
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4" Clamp x 1/2" ID; 1/Each
Cole-Parmer | Item: 5011810
Great for food, beverage, and dairy applications—fittings comply with FDA and 3-A standards
₹6,100.00INR / Each
₹6,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4" Clamp x 1/2" ID; 1/Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4" Clamp x 1/2" ID; 1/Each
5011810
₹6,100.00INR / Each
+ + + + + + + + +
Cole-Parmer Sanitary Gasket, Black FKM, 1/2"; 10/Pk
5013380
₹5,200.00INR / Pkg of 10
=
Total Price 94500.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Hose barb connection helps easily transition to tubing
  • 3/4" clamp size x 1/2" hose barb
  • 1 year warranty
More About this Item
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
1 Year Warranty
Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeSanitary x Hose Barb
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)1/2
  • Sanitary Clamp Size (Port 1) (InchShort)3/4
  • Flow PatternStraight
  • ColorMetallic
  • Regulatory Approvals3A;FDA
  • DescriptionFitting, 316L Stainless Steel, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4" Clamp x 1/2" ID