Cole-Parmer® Sanitary Gaskets, Black FKM
Compatible with Tri-Clamp® style fittings
  • Good steam and chemical resistance—suitable for SIP and CIP
  • Quick and easy gasket removal—gasket does not stick to the flanges
  • Exhibits excellent dimensional stability
  • Low extractable values
  • Usable temperature range: –34 to 204 °C (–30 to 400 °F)
  • Animal-derived ingredient free and phthalate free
  • FDA CFR 21 177 2600, USP Class VI, and 3A compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Made from high-quality fluorocarbon rubber material, these FKM gaskets provide exceptional chemical, temperature, and pressure resistance, making them suitable for SIP and CIP processes. FKM exhibits very low swell and minimal loss of physical properties after repeated steam cycling and long-term exposure to CIP media. Gaskets are free of phthalates, TSE/BSE, and animal-derived ingredients, ensuring the integrity of your fluid.
5,200.00 - ₹8,300.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sanitary Gasket Size (in) Seal Material Description
Availability Pricing
COMPARE
1/2 FKM Sanitary Gasket, Black FKM, 1/2"; 10/Pk
₹5,200.00INR / Pkg of 10
COMPARE
3/4 FKM Sanitary Gasket, Black FKM, 3/4"; 10/Pk
₹5,300.00INR / Pkg of 10
COMPARE
1 FKM Sanitary Gasket, Black FKM, 1"; 10/Pk
₹6,200.00INR / Pkg of 10
COMPARE
1 1/2 FKM Sanitary Gasket, Black FKM, 1-1/2"; 10/Pk
₹5,300.00INR / Pkg of 10
COMPARE
2 FKM Sanitary Gasket, Black FKM, 2"; 10/Pk
₹5,300.00INR / Pkg of 10
COMPARE
2 1/2 FKM Sanitary Gasket, Black FKM, 2-1/2"; 10/Pk
₹8,300.00INR / Pkg of 10