Cole-Parmer EX+ Adjustable Volume Pipette, 20-200 uL; Each
Representative image only.
Cole-Parmer EX+ Adjustable Volume Pipette, 20-200 uL; Each
Cole-Parmer | Item: 0791119
Chemical-resistant pipettor even handle solvents
₹72,900.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹72,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer EX+ Adjustable Volume Pipette, 20-200 uL; Each
Cole-Parmer EX+ Adjustable Volume Pipette, 20-200 uL; Each
0791119
₹72,900.00INR / Each
+
Cole-Parmer Fixed-Volume Pipettor, 1000 µL
2450116
₹5,400.00INR / Each
+ +
Cole-Parmer Pipette Tip, 20 to 200 µL, Bulk; 1000/BG
2450223
₹4,800.00INR / Bag of 1000
+
Cole-Parmer Pipette Tip, 100 to 1000 µL, Bulk; 1000/BG
2450261
₹5,700.00INR / Bag of 1000
=
Total Price 103300.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
calibration documentation.
Key Features
  • Fully autoclavable
  • UV resistant for flexible sterilization
  • One-touch calibration
  • Enhanced ergonomics
  • 2 year warranty
More About this Item
Chemical-resistant PTFE O-rings and seals allows these pipettors to be used with solvents. Fully autoclavable, adjustable volume pipettor features one-touch calibration and enhanced ergonomics. Enhanced UV resistance provides added protection from biohazardous elements. Easily adjust the pipette volume by turning the colored dial, and lock dial feature prevents accidental volume changes during use. Separate tip release button eliminates accidental release.
Two year warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)200
  • Min Volume (µL)20
  • Min Volume (mL)0.02
  • Max Volume (mL)0.2
  • Min Dispensing (mL)0.02
  • Max Dispensing (mL)0.2
  • Increments (µL)1
  • Accuracy (%)±1.00% to ±0.80%
  • Number Of Channels1
  • Tip EjectorYes
  • Body MaterialAl2O3 ceramic and PP plunger assembly
  • DescriptionAdjustable Volume Pipette, 20-200 uL; Each
  • Warranty2 Years