Cole-Parmer Pipette Tip, 20 to 200 µL, Sterile, Low Retention, Individually Wrapped; 400/PK
Representative image only.
Cole-Parmer Pipette Tip, 20 to 200 µL, Sterile, Low Retention, Individually Wrapped; 400/PK
Cole-Parmer | Item: 2450242
Tips free of human touch is purity at its best
₹8,600.00INR / Pkg of 400
₹8,600.00INR / Pkg of 400
Product Description
Key Features
  • Tip Options: Standard or low retention, sterile and combinations
  • Low retention tips free of any softening agents that could influence results of samples
  • Tips also leach-free, ensuring maximum recovery of sample yield
  • Volume measurements, along the tube are marked in increments for easy identification
  • Produced from Swiss-made Diamond Finish Mold
  • Made of FDA approved special medical grade virgin polypropylene plastic with high transparency
  • Unique label coding in packaging for easy identification
  • Sterile, low retention tips Volume: 20 to 200 µL
More About this Item
Not your everyday graduated pipette tips. It starts with a touch-free manufacturing facility and stringent quality control process to make these clear, plastic tips certified free of any affecting biological elements, such as human DNA, DNase and RNase and Endotoxin, or Pyrogen and PCR inhibitors. They also are noncytotoxic.
It’s about their innovative design, too. The profile and dimensions of the tips are engineered to accurately measure and transfer a volume of liquid from one container to another, as well as adapt to a wide-range of pipettes.
It ends with preserving their purity in the following packaging options, which are free of lubricants and heavy metals.
Individually Wrapped Tips: Free of DNase, RNase and Pyrogen
Bio DNase-FreeNonpyrogenicRNase-FreeSterile
Specifications
  • SterileYes
  • Max Volume (µL)200
  • Min Volume (µL)20
  • Tip StyleLow Retention, 5.34 cm L
  • Body MaterialPolypropylene (PP)
  • Filter TypeNone
  • Package OfIndividually Wrapped, 5 Packs of 80 Tips
  • DescriptionPipette Tip, 20 to 200 µL, Sterile, Low Retention, Individually Wrapped; 400/PK