Cole-Parmer Long Pipette Tip, 20 to 200 µL, Filter, 10 Racks; 960/PK
Representative image only.
Cole-Parmer Long Pipette Tip, 20 to 200 µL, Filter, 10 Racks; 960/PK
Cole-Parmer | Item: 2450251
Reach new depths, while at the same time, avoid the potential for carryover contamination
₹14,100.00INR / Pkg of 960
₹14,100.00INR / Pkg of 960
Buy It Together
Current Item
Cole-Parmer Long Pipette Tip, 20 to 200 µL, Filter, 10 Racks; 960/PK
Cole-Parmer Long Pipette Tip, 20 to 200 µL, Filter, 10 Racks; 960/PK
2450251
₹14,100.00INR / Pkg of 960
+
Cole-Parmer EX Adjustable Volume Pipette, 0.1-2.0 uL; Each
0790901
₹66,600.00INR / Each
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
=
Total Price 89100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Made in a touch-free manufacturing facility with a stringent control process
  • Reach the bottom of vessels without the pipette shaft touching its sides further protecting samples
  • Tip Options: Standard or low retention, filter and combinations
  • Graduation marks act as visual checks to lessen any risk of error, during sample aspiration
  • Produced from a Swiss-made Diamond Finish Mold
  • Made of FDA approved special medical grade virgin polypropylene plastic with high transparency
  • Multiple options for packaging: Bulk packs, racks and reloads
  • Packaging that is free of lubricants and heavy metals
  • Unique label coding in packaging for easy identification
  • Tips with filters Volume: 20 to 200 µL
More About this Item
These extended length clear pipette tips crafted of polypropylene feature a special design to prevent DNA denaturalization and, adapt with a wide-range of pipettes. Low retention tips free of all softening agents that could influence results of samples are offered. PE filter tips are specially designed to stop aerosols and contamination from entering pipettes, therefore, lessening the risk of cross contamination.
They are certified free of all affecting biological elements, such as human DNA, DNase and RNase and Endotoxin, or Pyrogen and PCR inhibitors, as well as heavy metals. The plastic tips also are noncytotoxic.
Specifications
  • SterileNo
  • Max Volume (µL)200
  • Min Volume (µL)20
  • Tip StyleStandard, 5.93 cm L
  • Body MaterialPolypropylene (PP)
  • Filter TypePolyethylene (PE)
  • Package OfStandard Racks, 10 x 96
  • DescriptionLong Pipette Tip, 20 to 200 µL, Filter, 10 Racks; 960/PK