Cole-Parmer Essentials EPA Precleaned TOC Vials, Glass with PTFE/Silicone Septa PP Cap; 80/Cs
Representative image only.
Cole-Parmer Essentials Item # WW-03756-20
Cleaned and certified for Total Organic Carbon (TOC) to <10ppb
  • Vials are constructed of clear borosilicate glass
₹56,200.00INR / Case of 80
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)40
  • Tube MaterialBorosilicate Glass
  • Cap MaterialPolypropylene
  • Septum TypePTFE/silicone
  • AutoclavableYes
  • Outside Diameter (in)1 1/10
  • Outside Diameter (mm)28
  • DescriptionEPA Precleaned TOC Vials, Glass with PTFE/Silicone Septa PP Cap; 80/Cs
Key Features
  • Vials are constructed of clear borosilicate glass
More About this Item
Vials are certified and traceable as associated with "Good Manufacturing Practices" (GMP) and include a Certificate of Analysis with every pack. Vials are manufactured from clear borosilicate glass and include attached white polypropylene hole cap and PTFE/silicone septa. They are packaged in an anti-static clotted plastic case that can be used in cleanroom applications.