Cole-Parmer Chromatography Tested 96-Well Plate, Polypropylene, Square Opening, U-Shape, 1600 µL; 96/Cs
Representative image only.
Cole-Parmer Chromatography Tested 96-Well Plate, Polypropylene, Square Opening, U-Shape, 1600 µL; 96/Cs
Cole-Parmer | Item: 6482402
Chromatography tested well plates are optimized for sample processing
₹151,000.00INR / Case of 96
₹151,000.00INR / Case of 96
Buy It Together
Current Item
Cole-Parmer Chromatography Tested 96-Well Plate, Polypropylene, Square Opening, U-Shape, 1600 µL; 96/Cs
Cole-Parmer Chromatography Tested 96-Well Plate, Polypropylene, Square Opening, U-Shape, 1600 µL; 96/Cs
6482402
₹151,000.00INR / Case of 96
+ =
Total Price 204300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Polypropylene microplates are manufactured from GC tested material
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • U- and V- bottom wells offer optimal sample recovery
  • Plate has square opening with U-shaped wells
More About this Item
The chromatography tested 96-well plates are ideal for pharmaceutical applications, sample collection and storage, combinatorial chemistry, and HTS applications. Plates meet SBS/ANSI standard footprint guidelines to assure compatibility with all well plate capable prep stations and chromatography autosamplers.
Specifications
  • Number of Wells96
  • Well Volume (µL)1600
  • Well ShapeSquare Opening, U-Shape
  • Surface TreatmentNone
  • SterileNo
  • Plate ColorClear
  • MaterialPolypropylene
  • Height (mm)31
  • DescriptionChromatography Tested 96-Well Plate, Polypropylene, Square Opening, U-Shape, 1600 µL; 96/Cs