Cole-Parmer Sealing Mat for Chromatography Tested 96-Well Plates, EVA, Square Well; 50/Cs
Representative image only.
Cole-Parmer Sealing Mat for Chromatography Tested 96-Well Plates, EVA, Square Well; 50/Cs
Cole-Parmer | Item: 6482406
Prevent spillage and contamination between wells
₹70,900.00INR / Case of 50
₹70,900.00INR / Case of 50
Product Description
Key Features
  • Tight seal reduces evaporation losses
  • Keep samples safe during transport or storage
  • EVA mat is suitable for use with square well openings
More About this Item
The chromatography tested 96-well sealing mats provide a secure seal for your 96-well plates, preventing evaporation loss, reducing cross contamination, and protecting samples. Sealing mats are available in round or square well shapes and a wide range of materials.
Specifications
  • Well ShapeSquare
  • Surface TreatmentNone
  • SterileNo
  • Plate ColorClear
  • MaterialEVA
  • DescriptionSealing Mat for Chromatography Tested 96-Well Plates, EVA, Square Well; 50/Cs