Cole-Parmer® Chromatography Tested Sealing Mats
Prevent spillage and contamination between wells
  • Tight seal reduces evaporation losses
  • Keep samples safe during transport or storage
The chromatography tested 96-well sealing mats provide a secure seal for your 96-well plates, preventing evaporation loss, reducing cross contamination, and protecting samples. Sealing mats are available in round or square well shapes and a wide range of materials.
12,400.00 - ₹55,600.00INR / Case of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Well Shape Description
Availability Pricing
COMPARE
Silicone Round Sealing Mat for Chromatography Tested 96-Well Plates, Silicone, Round 8 mm Diameter Well; 5/Cs
₹48,400.00INR / Case of 50
COMPARE
EVA Round Sealing Mat for Chromatography Tested 96-Well Plates, EVA, Round 8 mm Diameter Well; 5/Cs
₹12,400.00INR / Case of 5
COMPARE
Silicone/PTFE Round Sealing Mat for Chromatography Tested 96-Well Plates, Silicone/PTFE, Round 8 mm Diameter Well; 5/Cs
₹14,400.00INR / Case of 5
COMPARE
EVA Square Sealing Mat for Chromatography Tested 96-Well Plates, EVA, Square Well; 50/Cs
₹55,600.00INR / Case of 50
COMPARE
Silicone Square Sealing Mat for Chromatography Tested 96-Well Plates, Silicone, Square Well; 50/Cs
₹42,200.00INR / Case of 50