Cole-Parmer® Chromatography Tested 96-Well Plates
Chromatography tested well plates are optimized for sample processing
  • Polypropylene microplates are manufactured from GC tested material
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • U- and V- bottom wells offer optimal sample recovery
The chromatography tested 96-well plates are ideal for pharmaceutical applications, sample collection and storage, combinatorial chemistry, and HTS applications. Plates meet SBS/ANSI standard footprint guidelines to assure compatibility with all well plate capable prep stations and chromatography autosamplers.
7,400.00 - ₹151,000.00INR / Case of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Well Shape Well Volume (µL)
Availability Pricing
COMPARE
Polypropylene Round Opening, V-Shape 450
₹15,000.00INR / Case of 20
COMPARE
Polypropylene Round Opening, U-Shape 1100
₹75,100.00INR / Case of 50
COMPARE
Polypropylene Square Opening, V-Shape 2000
₹7,400.00INR / Case of 5
COMPARE
Polypropylene Square Opening, U-Shape 1600
₹151,000.00INR / Case of 96