Cole-Parmer BioSafe BioSafe Serological Pipettes, 10 mL; 200/Cs

Representative image only.

Cole-Parmer BioSafe BioSafe Serological Pipettes, 10 mL; 200/Cs

Cole-Parmer | Item: 1300253

Filtered for sample integrity and user safety

Buy It Together

Current Item

Cole-Parmer BioSafe BioSafe Serological Pipettes, 10 mL; 200/Cs
Cole-Parmer BioSafe BioSafe Serological Pipettes, 10 mL; 200/Cs
1300253

₹6,900.00INR / Case of 200
+
Cole-Parmer Acrylic Pipette Organizing Rack; Blue
0439507

₹23,900.00INR / Each
=
Total Price 30800.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Eliminate cross-contamination — unique barrier filter prevents over-aspiration
  • Polystyrene pipettes are sterile, individually wrapped, and disposable
  • Ideal for use with potentially hazardous or dangerous biological fluids
  • Use in place of filtered pipette tips

More About this Item

These polystyrene serological pipettes feature a special filter plug to lock in fluid and vapors if over aspiration occurs. They also protect electronic pipettors from contamination and cross-contamination of samples from over-aspiration — promoting user safety and sample integrity.

Specifications
  • SterileYes
  • AutoclavableNo
  • Max Volume (mL)10
  • Graduation Interval0.1 mL
  • Accuracy (%)±0.2 mL
  • Tip StyleStandard
  • Body MaterialPolystyrene
  • CalibrationTo deliver (TD)
  • DescriptionBioSafe Serological Pipettes, 10 mL; 200/Cs
Reviews
Q&A