Cole-Parmer Adjustable-Volume Pipettor, 0.1 to 2.5 µL
Representative image only.
Cole-Parmer Adjustable-Volume Pipettor, 0.1 to 2.5 µL
Cole-Parmer | Item: 2450101
Ergonomic Design Meets Precision in Liquid Handling
₹8,400.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹8,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Adjustable-Volume Pipettor, 0.1 to 2.5 µL
Cole-Parmer Adjustable-Volume Pipettor, 0.1 to 2.5 µL
2450101
₹8,400.00INR / Each
+
Cole-Parmer Pipette Tips, 1-10 mL, White; 200/Pk
0790925
₹14,000.00INR / Bag of 200
+ + +
Cole-Parmer Fixed-Volume Pipettor, 1000 µL
2450116
₹5,400.00INR / Each
+
Cole-Parmer Pipette Tip, 0.1 to 10 µL, Bulk; 1000/BG
2450142
₹4,500.00INR / Bag of 1000
+
Cole-Parmer Pipette Tip, 0.1 to 10 µL, 10 Racks; 960/PK
2450148
₹2,900.00INR / Pkg of 960
+
Cole-Parmer Long Pipette Tip, 20 to 200 µL, Bulk; 1000/BG
2450243
₹4,900.00INR / Bag of 1000
+
Cole-Parmer Pipette Tip, 100 to 1000 µL, Bulk; 1000/BG
2450261
₹5,700.00INR / Bag of 1000
+ =
Total Price 89400.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Color-coding drop and removal tool, calibration tool and silicon grease, along with sample tips and calibration report.
Key Features
  • Adjustable Volume – Easily set precise liquid volumes for accuracy
  • Ergonomic Design – Reduces hand strain for comfortable pipetting
  • Durable Construction – Built for long-term, high-precision use
  • Smooth Operation – Consistent plunger and tip ejection for efficiency
  • Universal Tip Compatibility – Works with most standard pipette tips
  • Volume: 0.1 to 2.5 µL
  • 3 year warranty
More About this Item
The Cole-Parmer® Single-Channel Adjustable Pipettor delivers precision, comfort, and durability for lab professionals. Its adjustable volume control allows for high-accuracy liquid transfers, while the ergonomic design reduces hand fatigue during extended use. The smooth plunger action and secure tip fit ensure consistent, contamination-free pipetting. Built for long-term reliability, this pipettor is an essential tool for research, diagnostics, and quality control.
3 Year WarrantyConformite EuropeenneISO 9001:2008 Certified Supplier
Specifications
  • AutoclavableYes
  • Max Volume (µL)2.5
  • Min Volume (µL)0.1
  • Increments (µL)0.01
  • Accuracy (%)2.5
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeGray
  • DescriptionAdjustable-Volume Pipettor, 0.1 to 2.5 µL; 1/EA