Cole-Parmer 3" Diameter Stainless Steel Sieve Cover

Representative image only.

Cole-Parmer 3" Diameter Stainless Steel Sieve Cover

Cole-Parmer | Item: 5998803

Protect and collect samples easily

₹6,900.00INR / Each

₹6,900.00INR / Each

Buy It Together

Current Item

Cole-Parmer 3" Diameter Stainless Steel Sieve Cover
Cole-Parmer 3" Diameter Stainless Steel Sieve Cover
5998803

₹6,900.00INR / Each
+ + + + =
Total Price 464500.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Suitable for use with 3” diameter sieves
  • Constructed of durable stainless steel
  • 1 year warranty

More About this Item

Use covers and receiving pans with 3” diameter sieves. Order sieves separately. Order a receiving pan for the bottom sieve to collect the finest particles.

Use a cover to protect your sample from contaminants.

1 Year Warranty
Specifications
  • MaterialStainless Steel
  • Description3" Diameter Stainless Steel Sieve Cover
  • Warranty1 Year
Reviews
Q&A