Carver 2090 Press Test Cylinder with Pellet Molds; 1-1/8" ID
Representative image only.
Carver 2090 Press Test Cylinder with Pellet Molds; 1-1/8" ID
Carver | Mfr: 2090| Item: 5962080
₹299,000.00INR / Each
₹299,000.00INR / Each
Buy It Together
Current Item
Carver 2090 Press Test Cylinder with Pellet Molds; 1-1/8" ID
Carver 2090 Press Test Cylinder with Pellet Molds; 1-1/8" ID
5962080
₹299,000.00INR / Each
+ =
Total Price 299000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Test Cylinders with Pellet Molds can be used for molding, tableting, crushing, extruding, or sample preparation. Stainless steel (SS) construction. Cylinders include base plunger, U-shaped ejector, SS receiving pan (3/4''D x 5-1/2'' dia.), and filters. Use pan and filters to collect sample residue to determine percentages. Order replacement filters separately.
1 Year Warranty
Specifications
  • Inside Diameter (in)1 1/8
  • DescriptionPress Test Cylinder with Pellet Molds; 1-1/8" ID