Carver Press Test Cylinders with Pellet Molds
Test Cylinders with Pellet Molds can be used for molding, tableting, crushing, extruding, or sample preparation. Stainless steel (SS) construction. Cylinders include base plunger, U-shaped ejector, SS receiving pan (3/4''D x 5-1/2'' dia.), and filters. Use pan and filters to collect sample residue to determine percentages. Order replacement filters separately.
299,000.00 - ₹450,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Inside Diameter (in)
Availability Pricing
Carver 2090 Press Test Cylinder with Pellet Molds; 1-1/8" ID 5962080 Mfr # 2090
COMPARE
1 1/8
₹299,000.00INR / Each
Carver 2091-5 Press Test Cylinder with Pellet Molds; 2-1/4" ID 5962082 Mfr # 2091-5
COMPARE
2 1/4
₹450,000.00INR / Each