Stainless steel cooling coil lets you operae below or slightly above ambient temperature
Temperature selection switch lets you quickly select from 10 popular temperature settings
Motor-driven stirrer ensures uniform temperature throughout bath
Ideal for kinematic viscosity measurements per ASTM D 445
1 year warranty
More About this Item
Control the viscosity of your samples with these Cannon® constant-temperature baths. These unique baths hold up to seven glass capillary viscometers and two thermometers (holders are required—order separately). See-through glass chamber lets you continuously view samples. All units feature a control circuit that provides proportional temperature control and an accuracy of ±0.01°>C from 20 to 100°>C (±0.03°>C above 100°>C for the high-temperature deluxe models). Select any temperature within the range or select from ten preset temperatures. Fine tuning dial lets you adjust this value to within 0.01°>C of your target temperature. Multiple safety features include overtemperature, low-level, and sensor disconnection cutoffs. Chamber is made of clear borosilicate glass and includes a reflective stainless steel baffle in the center; the deluxe model also includes two fluorescent lamps on both sides. Housing is constructed of corrosion-resistant epoxy-coated heavy aluminum with insulated stainless steel cover; includes stainless steel cooling coil. Fill bath with deionized water or oxidation-inhibiting bath oil for high-temperature applications. Order bath oil and other accessories separately, and thermometers from our Temperature section.
Specifications
Bath TypeViscosity Water Bath
Reservoir Capacity (L)10
Max Temperature (° C)100
Min Temperature (° C)20
Temperature Accuracy (° C)±0.01°C at 100°C
Low-Level CutoffYes
High-Temperature CutoffYes
Temperature SettingAnalog
Temperature SensorThermistor
Wetted MaterialsGlass and SS
Power (VAC)230
Descriptionstandard constant temperature bath, 240 VAC