Cannon 9726-M76 Brass viscometer holder for -Ubbelohde viscometers
Representative image only.
Cannon 9726-M76 Brass viscometer holder for -Ubbelohde viscometers
Cannon | Mfr: 9726-M76| Item: 9893494
Optimize performance of your Cannon Ubbelohde viscometers
₹63,600.00INR / Each
₹63,600.00INR / Each
Buy It Together
Current Item
Cannon 9726-M76 Brass viscometer holder for -Ubbelohde viscometers
Cannon 9726-M76 Brass viscometer holder for -Ubbelohde viscometers
9893494
₹63,600.00INR / Each
+
Cannon 9721-B65 Viscometer, universal size 200
9893455
₹49,400.00INR / Each
+
Cannon 9721-R59 Viscometer, Size 1; 2 to 10 Centistokes
9893413
₹75,900.00INR / Each
=
Total Price 188900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your viscometer running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your viscometers.
For holding Cannon Ubbelohde glass capillary viscometers in constant-temperature viscosity baths.
1 Year Warranty
Specifications
  • Viscometer TypeFor Cannon Ubbelohde
  • MaterialBrass
  • DescriptionBrass viscometer holder for -Ubbelohde viscometers