Cannon 9726-L20 Bath oil, 6 gallons (Accessories for Viscosity Baths)
Representative image only.
Cannon 9726-L20 Bath oil, 6 gallons (Accessories for Viscosity Baths)
Cannon | Mfr: 9726-L20| Item: 9893552
Optimize performance of your Cannon Ubbelohde viscometers
₹94,300.00INR / Each
₹94,300.00INR / Each
Buy It Together
Current Item
Cannon 9726-L20 Bath oil, 6 gallons (Accessories for Viscosity Baths)
Cannon 9726-L20 Bath oil, 6 gallons (Accessories for Viscosity Baths)
9893552
₹94,300.00INR / Each
+
Cannon 9721-B65 Viscometer, universal size 200
9893455
₹49,400.00INR / Each
+
Cannon 9721-R59 Viscometer, Size 1; 2 to 10 Centistokes
9893413
₹75,900.00INR / Each
=
Total Price 219600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your viscometer running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your viscometers.
Use bath oil for high-temperature baths. Viscosity is 36 cSt at 104°F (40°C); 5.6 cSt at 212°F (100°C). Use below 275°F (135°C).
1 Year Warranty
Specifications
  • DescriptionBath oil, 6 gallons (Accessories for Viscosity Baths)