Airfiltronix PF-1R PCR Prep Station Dacron Pre-Filter
Representative image only.
Airfiltronix PF-1R PCR Prep Station Dacron Pre-Filter
Airfiltronix | Mfr: PF-1R| Item: 4401115
A filter for every application
₹64,100.00INR / Pkg of 1
₹64,100.00INR / Pkg of 1
Buy It Together
Current Item
Airfiltronix PF-1R PCR Prep Station Dacron Pre-Filter
Airfiltronix PF-1R PCR Prep Station Dacron Pre-Filter
4401115
₹64,100.00INR / Pkg of 1
+ + + =
Total Price 1439100.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ensure optimal performance of your ductless fume hood
More About this Item
Choose from a wide range of filters to remove pollutants. These are compatible with the Aitfiltronix range of ductless fume hoods.
Specifications
  • Compatible WithAirfiltronix mini-ductless fume hoods 33705-41 and -42 and PCR prep station 44011-10
  • Length (InchShort)10 1/2
  • Width (InchShort)10 1/2
  • Depth (InchShort)1
  • TypeDacron Pre-Filter, 10.5"L x 10.5"W x 1" Deep
  • DescriptionDacron Pre-Filter, 10.5"L x 10.5"W x 1" Deep