Airfiltronix FRAM-4 Ammonia Gas Filter for Ductless Fume Hoods, 3/bx
Representative image only.
Airfiltronix FRAM-4 Ammonia Gas Filter for Ductless Fume Hoods, 3/bx
Airfiltronix | Mfr: FRAM-4| Item: 3370535
A filter for every application
₹292,000.00INR / Box of 3
₹292,000.00INR / Box of 3
Buy It Together
Current Item
Airfiltronix FRAM-4 Ammonia Gas Filter for Ductless Fume Hoods, 3/bx
Airfiltronix FRAM-4 Ammonia Gas Filter for Ductless Fume Hoods, 3/bx
3370535
₹292,000.00INR / Box of 3
+ + + =
Total Price 1667000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ensure optimal performance of your ductless fume hood
  • 1 year warranty
More About this Item
Choose from a wide range of filters to remove pollutants. These are compatible with the Aitfiltronix range of ductless fume hoods.
1 Year Warranty
Specifications
  • Compatible WithAirfiltronix ductless fume hoods 33705-16 to -22, ductless fume and particulate removal workstations 33705-02 to -08, portable fume/particle extractors 33705-43 to -46, and balance enclosure 44011-00
  • Length (InchShort)17
  • Width (InchShort)10
  • Depth (InchShort)2
  • TypeAmmonia Gas Specialty Carbon Filter
  • DescriptionAmmonia Gas Specialty Carbon Filter for Ductless Fume Hoods; 3/Box