Airfiltronix FRAG-4 Acid Gas Filter, 2" Deep; 3/Box
Representative image only.
Airfiltronix FRAG-4 Acid Gas Filter, 2" Deep; 3/Box
Airfiltronix | Mfr: FRAG-4| Item: 3370534
A filter for every application
₹292,000.00INR / Box of 3
₹292,000.00INR / Box of 3
Buy It Together
Current Item
Airfiltronix FRAG-4 Acid Gas Filter, 2" Deep; 3/Box
Airfiltronix FRAG-4 Acid Gas Filter, 2" Deep; 3/Box
3370534
₹292,000.00INR / Box of 3
+ + + =
Total Price 1667000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ensure optimal performance of your ductless fume hood
  • Ideal for ductless fume hoods
  • Breaks compounds down into the various elements for easy adsorption
  • 1 year warranty
More About this Item
Choose from a wide range of filters to remove pollutants. These are compatible with the Aitfiltronix range of ductless fume hoods.
1 Year Warranty
Specifications
  • Compatible WithAirfiltronix ductless fume hoods 33705-16 to -22, ductless fume and particulate removal workstations 33705-02 to -08, portable fume/particle extractors 33705-43 to -46, and balance enclosure 44011-00
  • Length (InchShort)17
  • Width (InchShort)10
  • Depth (InchShort)1/2
  • TypeAcid Gas Specialty Carbon Filter, 2" Deep
  • DescriptionAcid Gas Specialty Carbon Filter, 2" Deep; 3/Box