Traceable with NIST-Traceable Calibration
The Traceable® brand is synonymous with reliability and accuracy in measuring, monitoring and recording. These efficient, affordable instruments use current technology to simplify attaining the results you need to manage your critical process. Each Traceable product goes through an intense vetting process and comes with NIST-traceable certification. This means our accredited calibration testing lab has confirmed the product meets or exceeds all stated specifications. You can be certain the instrument provides accurate measurements right out of the box, saving both time and money by not having to calibrate separately. All NIST-traceable calibration certificates come from our ISO 17025 calibration laboratory with A2LA accreditation. This allows you to meet quality standards and regulatory compliance of ISO, FDA, cGMP, VFC, CAP, CLIA, as well as Joint Commission requirements. Select from thermometers, hygrometers, barometers, timers, clocks, stopwatches, meters, controllers, pumps, lab tools, and more.
There’s a Traceable solution for every level of monitoring
From basic measurement to cloud-based remote monitoring, there’s a Traceable product to fit every need.
Basic Monitoring
Monitor parameters with an easy-to-read digital display.
  • Accredited Calibration
  • Min/Max Monitoring
  • Hourly/Daily Recording
  • High/Low Alarms
Data Logger Monitoring
Automatically measure, monitor, and record parameters over a defined time period. View, download, and analyze collected data.
  • Log Data Continuously
  • Download Data to PC
  • Alarm History Reporting
  • Basic Monitoring Capabilities
Cloud Monitoring
Securely and wirelessly monitor critical environments and get alerts with TraceableLIVE®-capable products.
  • Real-Time Monitoring and Alerts
  • Receive Email, Text, or Push Notifications
  • Unlimited Data Cloud Storage
  • Built-In Reports—Includes VFC
  • Access/Activate via TraceableLIVE App or Traceablelive.com
TraceableLIVE
REMOTE MONITORING WITH TRACEABLELIVE
Everyone on your team can receive email, text, and push notifications on the environments you monitor.
Traceable Brands
Digi-Sense Traceable
Cole-Parmer Traceable
Related Articles
article
Articles and White Papers
Cole-Parmer NIST-Traceable Calibration and Certification
Let us provide you with the documentation you need to meet your most stringent quality requirements for the control of inspection, measuring, and test equipment..
article
Case Study
Why calibrate test equipment?
You’re serious about your electrical test instruments. You buy top brands, and you expect them to be accurate. You know some people send their digital instruments to a metrology lab for calibration, and you wonder why. After all, these are all electronic — there’s no meter movement to go out of balance. What do those calibration folks do, anyhow — just change the battery?